Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252501.1 | NP_001239430.1 | 21 | 3000 | ||
NM_001290356.1 | NP_001277285.1 | 21 | 3000 | |||
NM_021377.3 | NP_067352.2 | 21 | 3000 | |||
GenBank mRNA | AF137367.1 | - | 22 | 2836 | ||
AF195056.1 | - | 22 | 2836 | |||
AF284755.1 | - | 22 | 2836 | |||
AK030440.1 | - | 21 | 3001 | |||
AK046724.1 | - | 4 | 440 | |||
BC007486.1 | - | 13 | 1550 | 3590084 |