Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Pvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021424.2 | NP_067399.2 | 5 | 946 | ||
GenBank mRNA | AF239762.1 | - | 5 | 878 | ||
AF270977.1 | - | 5 | 878 | |||
AF297665.1 | - | 5 | 893 | |||
AK147479.1 | - | 1538 | ||||
BC060694.1 | - | 5 | 946 | 6405088 |