Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021434.5 | NP_067409.2 | 7 | 1130 | ||
GenBank mRNA | AB041545.1 | - | 7 | 1122 | ||
AK040165.1 | - | 1 | 26 | |||
AK053422.1 | - | 7 | 1100 | |||
AK169908.1 | - | 7 | 1129 | |||
AK170316.1 | - | 7 | 1095 | |||
BC055726.1 | - | 7 | 1105 | |||
BC069981.1 | - | 7 | 1086 | 6336270 |