Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021526.2 | NP_067501.2 | 12 | 1186 | ||
GenBank mRNA | AI448878.1 | - | 1 | 371 | ||
AK012013.1 | - | 12 | 1117 | |||
AK031863.1 | - | 12 | 1123 | |||
AK078134.1 | - | 12 | 1138 | |||
AK150280.1 | - | 12 | 1122 | |||
AK151104.1 | - | 12 | 1169 | |||
AK151733.1 | - | 12 | 1169 | |||
AK207350.1 | - | 1 | 31 | |||
BC003742.1 | - | 12 | 1091 | 3599671 | ||
Y13071.1 | - | 12 | 1107 |