Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021551.4 | NP_067526.2 | 9 | 1790 | ||
GenBank mRNA | AB012808.1 | - | 11 | 1576 | ||
AK007059.1 | - | 9 | 1735 | |||
AK029480.1 | - | 3 | 399 | |||
AK163116.1 | - | 3 | 3563 | |||
BC005727.1 | - | 7 | 2102 | |||
BC062878.1 | - | 9 | 1809 | 5696414 |