Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021555.2 | NP_067530.2 | 6 | 1110 | ||
GenBank mRNA | AB041552.1 | - | 6 | 1110 | ||
AK085815.1 | - | 6 | 1104 | |||
AK178524.1 | - | 4 | 391 | |||
BC061251.1 | - | 6 | 1074 | 30249656 |