Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101588.1 | NP_001095058.1 | 5 | 623 | ||
GenBank mRNA | BC158089.1 | - | 5 | 700 | 9088175 |