Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025336.1 | NP_079612.1 | 7 | 692 | ||
GenBank mRNA | AK002263.1 | - | 7 | 757 | ||
AK002889.1 | - | 7 | 763 | |||
AK003519.1 | - | 7 | 692 | |||
BC016528.1 | - | 6 | 514 | |||
BC021941.1 | - | 7 | 744 | 5124504 | ||
BC032987.1 | - | 7 | 707 |