Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025358.3 | NP_079634.2 | 7, 8 | 753 | ||
GenBank mRNA | AK003137.1 | - | 7, 8 | 735 | ||
AK003214.1 | - | 7, 8 | 726 | |||
AK012366.1 | - | 7, 8 | 733 | |||
BC005760.1 | - | 6, 7 | 634 | |||
BC058378.1 | - | 7, 8 | 721 | 6820787 |