Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025387.3 | NP_079663.1 | 3 | 253 | ||
GenBank mRNA | AK003891.1 | - | 3 | 202 | ||
AK008495.1 | - | 2 | 143 | |||
AK008878.1 | - | 3 | 175 | |||
AK010609.1 | - | 3 | 213 | |||
AK012101.1 | - | 3 | 200 | |||
AK019395.1 | - | 3 | 202 | |||
AK050688.1 | - | 3 | 188 | |||
AK089021.1 | - | 2 | 142 | |||
BC025854.1 | - | 3 | 154 | 5250659 | ||
BC055862.1 | - | 3 | 177 | 3492090 | ||
BC069899.1 | - | 2 | 899 | |||
BY785978.1 | - | 3 | 253 |