Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023203.1 | NP_075692.1 | 1, 2 | 156 | ||
GenBank mRNA | AF110764.1 | - | 1, 2 | 156 | ||
AK003643.1 | - | 1, 2 | 135 | |||
AK010508.1 | - | 1, 2 | 150 | |||
AK010606.1 | - | 1, 2 | 123 | |||
BC004623.1 | - | 1, 2 | 183 | 3585680 |