Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a6os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007567.2 | NP_001007568.1 | 3 | 604 | ||
GenBank mRNA | AK010254.1 | - | 3 | 591 | ||
AK134626.1 | - | 3 | 505 | |||
AK146733.1 | - | 3 | 605 | |||
AK171806.1 | - | 2 | 340 | |||
BC054118.1 | - | 3 | 602 | 30021749 |