Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025567.2 | NP_079843.1 | 2 | 218 | ||
GenBank mRNA | AK002815.1 | - | 2 | 196 | ||
AK011288.1 | - | 2 | 219 | |||
AK151299.1 | - | 2 | 209 | |||
AK170272.1 | - | 1 | 818 | |||
AK192516.1 | - | 2 | 74 | |||
BC005620.1 | - | 2 | 199 | 3711323 | ||
BC082790.1 | - | 2 | 156 |