Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usmg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023211.4 | NP_075700.2 | 2 | 128 | ||
GenBank mRNA | AK008345.1 | - | 2 | 129 | ||
AK008511.1 | - | 958 | ||||
AK038457.1 | - | 2 | 135 | |||
AK088856.1 | - | 2 | 146 | |||
BC024355.1 | - | 2 | 209 | |||
BC087919.1 | - | 2 | 128 | 6545418 |