Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps19bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175109.3 | NP_780318.1 | 2 | 200 | ||
GenBank mRNA | AB233218.1 | - | 2 | 196 | ||
AK010954.1 | - | 2 | 184 | |||
AK075974.1 | - | 2 | 181 | |||
BC038691.1 | - | 2 | 186 | 1495043 | ||
BC050115.1 | - | 2 | 189 | 30017673 | ||
BC065381.1 | - | 1 | 133 | |||
BY237968.1 | - | 2 | 200 |