Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025637.3 | NP_079913.2 | 2 | 217 | ||
GenBank mRNA | AK011007.1 | - | 2 | 217 | ||
AK011008.1 | - | 2 | 217 | |||
AK050254.1 | - | 1 | 84 | |||
BC128288.1 | - | 2 | 216 | |||
BC150838.1 | - | 2 | 217 | 9087749 | ||
BY710389.1 | - | 2 | 217 |