Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159609.1 | NP_001153081.1 | 5 | 769 | ||
NM_001159610.1 | NP_001153082.1 | 5 | 820 | |||
NM_001159612.1 | NP_001153084.1 | 5 | 836 | |||
NM_025657.3 | NP_079933.2 | 5 | 719 | |||
GenBank mRNA | AK003037.1 | - | 5 | 626 | ||
AK003592.1 | - | 5 | 737 | |||
AK012637.1 | - | 5 | 834 | |||
AK015684.1 | - | 5 | 771 | |||
AK019566.1 | - | 5 | 831 | |||
AK033084.1 | - | 1 | 3301 | |||
AK134971.1 | - | 5 | 678 | |||
AK169249.1 | - | 5 | 743 | |||
BC034894.1 | - | 5 | 678 | 3328402 |