Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080969.3 | NP_001074438.1 | 3, 4 | 581 | ||
NM_001290737.1 | NP_001277666.1 | 4, 5 | 524 | |||
GenBank mRNA | AK007233.1 | - | 5, 6 | 562 | ||
AK010876.1 | - | 3, 4 | 573 | |||
AK015467.1 | - | 3, 4 | 375 | |||
AK016076.1 | - | 4, 5 | 525 | |||
AK017532.1 | - | 3, 4 | 564 | |||
BC092541.1 | - | 3, 4 | 561 | 30280958 | ||
BC115560.1 | - | 3, 4 | 569 | 40101901 | ||
BC115561.1 | - | 3, 4 | 569 | 40101902 |