Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dcbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025705.3 | NP_079981.2 | 3, 4 | 559 | ||
GenBank mRNA | AK014521.1 | - | 3, 4 | 371 | ||
AK016485.1 | - | 3, 4 | 559 | |||
BC026771.1 | - | 2, 3 | 347 | 4014778 | ||
BY096153.1 | - | 2, 3 | 297 |