Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025718.3 | NP_079994.2 | 8 | 934 | ||
GenBank mRNA | AK003619.1 | - | 8 | 935 | ||
AK014633.1 | - | 7 | 1126 | |||
BC106188.1 | - | 8 | 898 | 30135288 |