Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025785.2 | NP_080061.1 | 10 | 1118 | ||
GenBank mRNA | AK008027.1 | - | 10 | 1122 | ||
AK018626.1 | - | 10 | 1119 | |||
AK134722.1 | - | 11 | 1146 | |||
AK188178.1 | - | 3 | 247 | |||
BC014749.1 | - | 10 | 1104 | 4240953 |