Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF373710.1 | - | 3, 4 | 181 | ||
AK003572.2 | - | 3, 4 | 211 | |||
AK003699.1 | - | 3, 4 | 135 | |||
AK085669.1 | - | 3, 4 | 260 | |||
BC013535.1 | - | 3, 4 | 144 | 4239189 | ||
BY083019.1 | - | 3, 4 | 193 |