Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025849.3 | NP_080125.1 | 2 | 353 | ||
GenBank mRNA | AK003640.1 | - | 2 | 324 | ||
AK006432.1 | - | 2 | 280 | |||
AK013937.1 | - | 2 | 356 | |||
BC022621.1 | - | 2 | 377 | 4192355 |