Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025870.1 | NP_080146.1 | 2 | 354 | ||
GenBank mRNA | AK009864.1 | - | 2 | 346 | ||
AK043948.1 | - | 2 | 323 | |||
BC019749.1 | - | 2 | 337 | |||
BC150493.1 | - | 2 | 356 | 40142491 |