Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc25a53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082412.2 | NP_001075881.1 | 7 | 1021 | ||
NM_001114179.1 | NP_001107651.1 | 2 | 403 | |||
GenBank mRNA | AK076279.1 | - | 7 | 1016 | ||
CX240158.1 | - | 2 | 403 |