Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026056.4 | NP_080332.1 | 10 | 1327 | ||
GenBank mRNA | AK010235.1 | - | 10 | 1074 | ||
AK013331.1 | - | 10 | 1084 | |||
AK014979.1 | - | 6 | 722 | |||
AK033071.1 | - | 10 | 1083 | |||
AK035556.1 | - | 10 | 1328 | |||
AK047701.1 | - | 8 | 894 | |||
AK049813.1 | - | 7 | 770 | |||
AK076737.1 | - | 6 | 747 | |||
BC050752.1 | - | 10 | 1088 | 6533812 | ||
BC057937.1 | - | 10 | 1142 | 5325599 |