Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026175.5 | NP_080451.4 | 11 | 1705 | ||
GenBank mRNA | AK004763.1 | - | 11 | 1688 | ||
AK028829.1 | - | 11 | 1657 | |||
AK029095.1 | - | 11 | 1675 | |||
AK030115.1 | - | 3 | 230 | |||
AK030223.1 | - | 11 | 1692 | |||
AK134178.1 | - | 11 | 1683 | |||
AK160061.1 | - | 11 | 1688 | |||
AK182650.1 | - | 2 | 162 | |||
BC010727.1 | - | 2 | 160 | |||
BC029753.1 | - | 11 | 1669 | 2651773 |