Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026211.3 | NP_080487.2 | 1 | 149 | ||
GenBank mRNA | AK010259.1 | - | 1 | 123 | ||
AK152496.1 | - | 1 | 142 | |||
AK152581.1 | - | 1 | 142 | |||
AK153383.1 | - | 1 | 142 | |||
AK169001.1 | - | 1 | 122 | |||
BC004691.1 | - | 1 | 133 | 3501295 | ||
BC049282.1 | - | 1 | 115 | 2648375 | ||
BC058801.1 | - | 1 | 115 | 6476565 |