Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026217.3 | NP_080493.2 | 2 | 279 | ||
GenBank mRNA | AB066216.1 | - | 2 | 251 | ||
AK005405.1 | - | 2 | 283 | |||
AK008698.1 | - | 2 | 255 | |||
AK016474.2 | - | 2 | 258 | |||
AK039546.1 | - | 2 | 385 | |||
AK167027.1 | - | 2 | 257 | |||
BB853539.1 | - | 2 | 280 | |||
BC070470.1 | - | 2 | 249 | 30630196 |