Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026217.3 | NP_080493.2 | 3 | 379 | ||
GenBank mRNA | AB066216.1 | - | 3 | 351 | ||
AK005405.1 | - | 3 | 383 | |||
AK008698.1 | - | 3 | 355 | |||
AK016474.2 | - | 3 | 358 | |||
AK167027.1 | - | 3 | 357 | |||
AK198454.1 | - | 2 | 109 | |||
AK208391.1 | - | 2 | 109 | |||
AK214439.1 | - | 2 | 120 | |||
BB853539.1 | - | 3 | 380 | |||
BC070470.1 | - | 3 | 349 | 30630196 |