Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930548H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026296.3 | NP_080572.1 | 2 | 744 | ||
GenBank mRNA | AK016690.1 | - | 2 | 745 | ||
BC057946.1 | - | 1 | 1840 |