Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146342.2 | NP_001139814.2 | 2 | 469 | ||
NM_001167818.1 | NP_001161290.1 | 4 | 718 | |||
GenBank mRNA | AK014866.1 | - | 4 | 587 | ||
AK170321.1 | - | 2 | 470 | |||
BC052355.1 | - | 4 | 549 | 6770333 | ||
DQ788834.1 | - | 4 | 495 |