Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026454.3 | NP_080730.1 | 3, 4 | 272 | ||
GenBank mRNA | AK007937.1 | - | 3, 4 | 272 | ||
AK007960.1 | - | 3, 4 | 262 | |||
AK010957.1 | - | 4, 5 | 349 | |||
AK030594.1 | - | 252 | ||||
AK153481.1 | - | 3, 4 | 273 | |||
BC016117.1 | - | 4, 5 | 554 | 4486610 |