Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182991.2 | NP_892036.1 | 4 | 569 | ||
GenBank mRNA | AK005155.1 | - | 4 | 533 | ||
BC056172.1 | - | 4 | 569 | 5699902 |