Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026503.3 | NP_080779.2 | 2 | 229 | ||
GenBank mRNA | AK003789.1 | - | 2 | 187 | ||
AK004301.1 | - | 2 | 209 | |||
AK007909.1 | - | 2 | 222 | |||
AK015824.1 | - | 2 | 230 | |||
BC028981.1 | - | 2 | 194 | 5367409 | ||
BF147474.1 | - | 2 | 443 |