Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700062C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026529.4 | NP_080805.1 | 3 | 151 | ||
GenBank mRNA | AK011228.1 | - | 2 | 238 | ||
AK012467.1 | - | 2 | 253 | |||
AK013030.1 | - | 3 | 61 | |||
AK018391.1 | - | 3 | 155 | |||
AK164298.1 | - | 1 | 1854 | |||
BC029246.1 | - | 3 | 156 | |||
BC037075.1 | - | 3 | 199 | 5038294 | ||
BC084681.1 | - | 3 | 161 | 30144888 |