Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133831.3 | NP_598592.2 | 4 | 620 | ||
GenBank mRNA | AK077341.1 | - | 4 | 574 | ||
AK079167.1 | - | 4 | 574 | |||
AK088461.1 | - | 4 | 577 | |||
AK146916.1 | - | 4 | 589 | |||
AK151836.1 | - | 4 | 577 | |||
AK158665.1 | - | 4 | 520 | |||
AK160070.1 | - | 4 | 576 | |||
AK169290.1 | - | 4 | 580 | |||
AY535001.1 | - | 4 | 565 | |||
BC017637.1 | - | 4 | 596 | 4912415 | ||
BC025810.1 | - | 4 | 527 |