Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168471.1 | NP_001161943.1 | 2 | 134 | ||
NM_001168472.1 | NP_001161944.1 | 2 | 307 | |||
NM_026556.4 | NP_080832.1 | 2 | 340 | |||
GenBank mRNA | AK006876.1 | - | 2 | 116 | ||
AK011284.1 | - | 2 | 308 | |||
AK028319.1 | - | 2 | 105 | |||
AK049645.1 | - | 2 | 314 | |||
AK078434.1 | - | 2 | 313 | |||
AK160114.1 | - | 2 | 310 | |||
AK169212.1 | - | 2 | 313 | |||
AY029255.1 | - | 1 | 42 | |||
BC011289.1 | - | 2 | 131 | 3981362 | ||
BC040822.1 | - | 2 | 302 | 4459858 | ||
BC049621.1 | - | 113 | ||||
BC058797.1 | - | 2 | 272 | |||
BY249698.1 | - | 2 | 307 | |||
BY310878.1 | - | 2 | 341 |