Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168471.1 | NP_001161943.1 | 2 | 201 | ||
NM_001168472.1 | NP_001161944.1 | 2 | 374 | |||
NM_026556.4 | NP_080832.1 | 2 | 407 | |||
GenBank mRNA | AK006876.1 | - | 2 | 183 | ||
AK011284.1 | - | 2 | 375 | |||
AK028319.1 | - | 2 | 172 | |||
AK049645.1 | - | 2 | 381 | |||
AK078434.1 | - | 2 | 380 | |||
AK160114.1 | - | 2 | 377 | |||
AK169212.1 | - | 2 | 380 | |||
AY029255.1 | - | 1 | 109 | |||
BC011289.1 | - | 2 | 198 | 3981362 | ||
BC040822.1 | - | 2 | 369 | 4459858 | ||
BC049621.1 | - | 180 | ||||
BY249698.1 | - | 2 | 374 |