Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fahd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029629.2 | NP_083905.1 | 2 | 236 | ||
GenBank mRNA | AK005134.1 | - | 2 | 237 | ||
AK080868.1 | - | 2 | 222 | |||
AK170875.1 | - | 2 | 210 | |||
BC030182.1 | - | 2 | 187 | 5044017 | ||
BY269533.1 | - | 2 | 237 |