Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam135a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026604.4 | NP_080880.4 | 14 | 3242 | ||
GenBank mRNA | AK012197.1 | - | 1 | 727 | ||
AK019549.1 | - | 1 | 860 | |||
AK034175.1 | - | 1 | 1608 | |||
AK044359.1 | - | 1 | 1406 | |||
AK173166.1 | - | 1 | 366 | |||
AK195972.1 | - | 1 | 111 | |||
BC031160.1 | - | 1 | 1264 | |||
BC066812.1 | - | 1 | 130 | |||
BC070446.1 | - | 13 | 2924 | 30362784 | ||
BC083093.1 | - | 1 | 721 |