Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026612.3 | NP_080888.1 | 2 | 202 | ||
GenBank mRNA | AK007495.1 | - | 2 | 178 | ||
AK010642.1 | - | 2 | 209 | |||
AK019008.1 | - | 2 | 207 | |||
AK028334.1 | - | 211 | ||||
AK216480.1 | - | 1 | 17 | |||
BC013510.1 | - | 2 | 212 | 4225177 |