Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026623.3 | NP_080899.1 | 2 | 325 | ||
GenBank mRNA | AK011688.2 | - | 2 | 315 | ||
AK012308.1 | - | 2 | 299 | |||
AK019433.1 | - | 2 | 308 | |||
AK020150.1 | - | 2 | 305 | |||
AK146419.1 | - | 2 | 291 | |||
AK147061.1 | - | 308 | ||||
AK160147.1 | - | 2 | 291 | |||
AK204985.1 | - | 2 | 205 | |||
AK217354.1 | - | 1 | 57 | |||
BC008270.1 | - | 2 | 308 | 2811199 | ||
BC090834.1 | - | 284 | 6855233 |