Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam96b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026753.2 | NP_081029.1 | 3 | 335 | ||
GenBank mRNA | AK003830.1 | - | 3 | 338 | ||
BC055880.1 | - | 3 | 309 | 4190754 |