Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277860.1 | NP_001264789.1 | 3 | 213 | ||
NM_001277861.1 | NP_001264790.1 | 3 | 227 | |||
NM_026798.3 | NP_081074.1 | 3 | 286 | |||
GenBank mRNA | AF332866.1 | - | 2 | 152 | ||
AK003740.1 | - | 3 | 288 | |||
AK013286.1 | - | 3 | 217 | |||
AK080725.1 | - | 1 | 1177 | |||
BC052053.1 | - | 3 | 190 | 5693205 | ||
BY127975.1 | - | 3 | 216 | |||
BY704000.1 | - | 3 | 288 | |||
DV063021.1 | - | 3 | 281 |