Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026817.3 | NP_081093.2 | 7 | 626 | ||
GenBank mRNA | AK004012.1 | - | 6 | 842 | ||
AK170205.1 | - | 4 | 314 | |||
AK180232.1 | - | 106 | ||||
AK209433.1 | - | 2 | 105 | |||
BC030376.1 | - | 7 | 593 | 5374611 | ||
BQ887131.1 | - | 7 | 618 |