Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110032F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167996.1 | NP_001161468.1 | 1 | 410 | ||
GenBank mRNA | AK082199.1 | - | 1 | 155 |