Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081185.1 | NP_001074654.1 | 45 | 7828 | ||
GenBank mRNA | AK135523.1 | - | 10 | 1668 | ||
BC039885.1 | - | 10 | 1696 | |||
BC052186.1 | - | 18 | 2777 | |||
BC060276.1 | - | 24 | 3889 | |||
BC151097.1 | - | 45 | 7633 | 9088010 | ||
BC158128.1 | - | 44 | 7534 | 9007396 | ||
BC172159.1 | - | 44 | 7534 |