Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081119.1 | NP_001074588.1 | 2 | 1245 | ||
NM_026868.1 | NP_081144.1 | 3 | 1352 | |||
GenBank mRNA | AK004375.1 | - | 1 | 435 | ||
AK044407.1 | - | 2 | 1099 | |||
AK215187.1 | - | 1 | 67 | |||
BC043690.1 | - | 2 | 1058 | 5148187 |