Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286217.1 | NP_001273146.1 | 5 | 390 | ||
NM_001286218.1 | NP_001273147.1 | 4 | 319 | |||
NM_026928.3 | NP_081204.2 | 5 | 390 | |||
GenBank mRNA | AA403787.1 | - | 4 | 250 | ||
AK007494.1 | - | 5 | 354 | |||
AK042452.1 | - | 353 | ||||
AK044731.1 | - | 5 | 355 | |||
AK163708.1 | - | 5 | 502 | |||
BC028662.1 | - | 5 | 351 | 1332340 | ||
CK618091.1 | - | 5 | 325 |